Silver trifluoromethanesulphonate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Silver trifluoromethanesulphonate, Silver triflate, Trifluoromethanesulphonic acid silver salt
M-Clarity™ quality level = MQ100
85325-25G
85325-5G
SIAL85325-25GEA
7930
CZK
SIAL85325-25G
SIAL85325-5G
Silver trifluoromethanesulphonate, Sigma-Aldrich®
Silver trifluoromethanesulphonate
Vzorec:
CAgF₃O₃S MW: 256,94 g/mol Bod tání: 356 °C |
MDL Number:
MFCD00013226 CAS číslo: 2923-28-6 EINECS: 220-882-2 UN: 3260 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...