Silver trifluoromethanesulphonate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Silver trifluoromethanesulphonate, Silver triflate, Trifluoromethanesulphonic acid silver salt
M-Clarity™ quality level = MQ100
176435-10G
176435-1G
176435-25G
SIAL176435-10GEA
3180
CZK
SIAL176435-10G
SIAL176435-1G
SIAL176435-25G
Silver trifluoromethanesulphonate, Sigma-Aldrich®
Silver trifluoromethanesulphonate
Vzorec:
CAgF₃O₃S MW: 256,94 g/mol Bod tání: 356 °C |
MDL Number:
MFCD00013226 CAS číslo: 2923-28-6 EINECS: 220-882-2 UN: 3260 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...