Tisk…

Silver trifluoromethanesulphonate ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Silver trifluoromethanesulphonate, Silver triflate, Trifluoromethanesulphonic acid silver salt

88722.04 88722.09 88722.18 AB205192.0050
88722.04EA 1310 CZK
88722.04 88722.09 88722.18 ABCRAB205192.0050
Silver trifluoromethanesulphonate ≥98%
Silver trifluoromethanesulphonate

Purity: 98% CAS No.: 2923-28-6 Molecular Formula: CAgF3O3S Formula Weight: 256.93 Physical Form: Crystalline Sensitivity: Light Sensitive Appearance: White to grey Odour: Odourless Melting Point: 356° UN No.: 3260 MDL No.: MFCD00013226

Vzorec: CAgF₃O₃S
MW: 256,94 g/mol
Bod tání: 356 °C
Storage Temperature: Ambient
MDL Number: MFCD00013226
CAS číslo: 2923-28-6
EINECS: 220-882-2
UN: 3260
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR