Tisk…

Silver trifluoromethanesulphonate 99+%

Dodavatel: Thermo Scientific
Varování

Synonyms: Silver trifluoromethanesulphonate, Silver triflate, Trifluoromethanesulphonic acid silver salt

175200100 175200250 175201000
ACRO175200100EA 5040 CZK
ACRO175200100 ACRO175200250 ACRO175201000
Silver trifluoromethanesulphonate 99+%
Silver trifluoromethanesulphonate
Vzorec: CAgF₃O₃S
MW: 256,94 g/mol
Bod tání: 356 °C
Storage Temperature: Ambient
MDL Number: MFCD00013226
CAS číslo: 2923-28-6
EINECS: 220-882-2
ADR: 8,III

Objednat

Výsledky specifikačního testu

Appearance White to light beige Powder
Infrared spectrum Conforms
Elemental Analysis ≥99.0 %
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR