Sodium trifluoromethanesulphonate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Trifluoromethanesulphonic acid sodium salt
M-Clarity™ quality level = MQ100
367907-25G
367907-5G
SIAL367907-25GEA
6670
CZK
SIAL367907-25G
SIAL367907-5G
Sodium trifluoromethanesulphonate, Sigma-Aldrich®
Sodium trifluoromethanesulphonate
Vzorec:
CF₃NaO₃S MW: 172,06 g/mol |
MDL Number:
MFCD00061607 CAS číslo: 2926-30-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...