Trifluoromethanesulphonic anhydride, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Triflic anhydride
M-Clarity™ quality level = MQ200
176176-10G
176176-1KG
176176-250G
176176-50G
176176-5G
SIAL176176-10GEA
1840
CZK
SIAL176176-10G
SIAL176176-1KG
SIAL176176-250G
SIAL176176-50G
SIAL176176-5G
Trifluoromethanesulphonic anhydride, Sigma-Aldrich®
Trifluoromethanesulphonic anhydride
Vzorec:
C₂F₆O₅S₂ MW: 282,14 g/mol Bod varu: 83 °C (1013 hPa) Hustota: 1,67 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000408 CAS číslo: 358-23-6 EINECS: 206-616-8 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...