Trifluoromethanesulphonic anhydride ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Triflic anhydride
A11767.22
A11767.14
A11767.09
A11767.18
A11767.30
A11767.22EA
14830
CZK
A11767.22
A11767.14
A11767.09
A11767.18
A11767.30
Trifluoromethanesulphonic anhydride ≥98%
Trifluoromethanesulphonic anhydride
Purity: 98%
CAS No.: 358-23-6
Molecular Formula: C2F6O5S2
Formula Weight: 282.13
Refractive Index: 1.3210
Melting Point: -80°
Boiling Point: 81-83°
Beilstein Reference: 1813600
UN No.: 3265
MDL No.: MFCD00000408
Vzorec:
C₂F₆O₅S₂ MW: 282,14 g/mol Bod varu: 81…83 °C Bod tání: –80 °C Hustota: 1,720 Storage Temperature: Ambient |
MDL Number:
MFCD00000408 CAS číslo: 358-23-6 EINECS: 206-616-8 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...