Trifluoromethanesulphonic anhydride 98+%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Triflic anhydride
175060010
175060500
175062500
ACRO175060010EA
1280
CZK
ACRO175060010
ACRO175060500
ACRO175062500
Trifluoromethanesulphonic anhydride 98+%
Trifluoromethanesulphonic anhydride
Vzorec:
C₂F₆O₅S₂ Bod varu: 80 °C (760 mmHg) Hustota: 1,7 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00000408 CAS číslo: 358-23-6 EINECS: 206-616-8 UN: 3265 ADR: 8,I |
Výsledky specifikačního testu
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3200 to 1.3230 (20°C, 589 nm) |
Titration with CHA | 98.0 to 101.0 % |
Impurity | ≤1.0 % CF3SO3H |
Distillation range | ≥95 % (between 123-133°C) |
Pour point | < -40°C |
Residue | ≤10 ug/ml |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...