Triisopropylsilyl trifluoromethanesulphonate ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Trifluoromethanesulphonic acid triisopropylsilyl ester, Tri(propan-2-yl)silyl trifluoromethanesulfonate
B21127.06
B21127.14
B21127.22
B21127.06EA
1760
CZK
B21127.06
B21127.14
B21127.22
Triisopropylsilyl trifluoromethanesulphonate ≥97%
Triisopropylsilyl trifluoromethanesulphonate
Purity: 97%
CAS No.: 80522-42-5
Molecular Formula: CF3SO3Si[CH(CH3)2]3
Formula Weight: 306.42
Flash Point: 100°(212°F)
Refractive Index: 1.4150
Boiling Point: 45-46°/0.03mm
Beilstein Reference: 3591541
UN No.: 3265
MDL No.: MFCD00009913
Vzorec:
C₁₀H₂₁F₃O₃SSi MW: 306,42 g/mol Bod varu: 45…46 °C (0,03 mmHg) Hustota: 1,136 Bod vzplanutí: 100 °C (212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009913 CAS číslo: 80522-42-5 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...