Tisk…

2,2,3,3,4,4,4-Heptafluoro-1-butanol, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: 1H,1H-Perfluoro-1-butanol, 2,2,3,3,4,4,4-Heptafluorobutan-1-ol, Heptafluoro-1-butanol

M-Clarity™ quality level = MQ200

H1604-25G H1604-5G
SIALH1604-25GEA 8500 CZK
SIALH1604-25G SIALH1604-5G
2,2,3,3,4,4,4-Heptafluoro-1-butanol, Sigma-Aldrich®
2,2,3,3,4,4,4-Heptafluoro-1-butanol
Vzorec: C₄H₃F₇O
MW: 200,06 g/mol
Bod varu: 96 °C (1013 hPa)
Hustota: 1,6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004674
CAS číslo: 375-01-9
EINECS: 206-782-1
UN: 1987
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR