Pentafluoropropionic anhydride, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Perfluoropropanoic anhydride, Pentafluoropropanoic anhydride, PFPA
77292-25ML
77292-5ML
SIAL77292-25MLEA
11400
CZK
SIAL77292-25ML
SIAL77292-5ML
Pentafluoropropionic anhydride, Sigma-Aldrich®
Pentafluoropropionic anhydride
Vzorec:
C₆F₁₀O₃ MW: 310,05 g/mol Bod varu: 69 °C (1013 hPa) Hustota: 1,571 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000429 CAS číslo: 356-42-3 EINECS: 206-604-2 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...