1-Iodoperfluorbutane, Sigma-Aldrich®
Dodavatel: Merck
|
|
Synonyms:
Perfluorobutyl iodide, 1,1,1,2,2,3,3,4,4-Nonafluoro-4-iodobutane, 1-Iodononafluorobutane, Nonafluoro-1-iodobutane
M-Clarity™ quality level = MQ100
317845-100G
317845-25G
SIAL317845-100GEA
5820
CZK
SIAL317845-100G
SIAL317845-25G
1-Iodoperfluorbutane, Sigma-Aldrich®
1-Iodoperfluorbutane
Vzorec:
C₄F₉I MW: 345,93 g/mol Bod varu: 65 °C (1013 hPa) Bod tání: –2 °C Hustota: 2,0497 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001062 CAS číslo: 423-39-2 EINECS: 207-025-8 UN: 3082 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...