1-Iodo-3,3,3-trifluoropropane ≥98%
Dodavatel: Thermo Scientific
Synonyms:
1,1,1-Trifluoro-3-iodopropane, 3-Iodo-1,1,1-trifluoropropane
B23284.03
B23284.06
B23284.03EA
920
CZK
B23284.03
B23284.06
1-Iodo-3,3,3-trifluoropropane ≥98%
1-Iodo-3,3,3-trifluoropropane
Purity: 98+%
CAS No.: 460-37-7
Molecular Formula: CF3CH2CH2I
Formula Weight: 223.96
Flash Point: None
Refractive Index: 1.4200
Boiling Point: 89-90°
Beilstein Reference: 1698182
MDL No.: MFCD00038531
Vzorec:
C₃H₄F₃I MW: 223,96 g/mol Bod varu: 89…90 °C Hustota: 1,911 Storage Temperature: Ambient |
MDL Number:
MFCD00038531 CAS číslo: 460-37-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...