Tetrafluorosuccinic acid ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
2,2,3,3-Tetrafluorosuccinic acid, Tetrafluoro-1,4-butanedioic acid
B20462.03
B20462.06
B20462.03EA
841
CZK
B20462.03
B20462.06
Tetrafluorosuccinic acid ≥97%
Tetrafluorosuccinic acid
Purity: 97%
CAS No.: 377-38-8
Molecular Formula: C4H2F4O4
Formula Weight: 190.05
Melting Point: ca 115°
Boiling Point: 150°/15mm
UN No.: 3261
MDL No.: MFCD00042430
Vzorec:
C₄H₂F₄O₄ MW: 190,05 g/mol Bod varu: 150 °C (15 mmHg) Bod tání: ∼ 115 °C Hustota: 1,400 Storage Temperature: Ambient |
MDL Number:
MFCD00042430 CAS číslo: 377-38-8 EINECS: 206-820-7 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...