Tisk…

Thallium(III) trifluoroacetate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

150533-10G 150533-50G 150533-5G
SIAL150533-10GEA 3870 CZK
SIAL150533-10G SIAL150533-50G SIAL150533-5G
Thallium(III) trifluoroacetate, Sigma-Aldrich®
Thallium(III) trifluoroacetate
Vzorec: C₆F₉O₆Tl
Bod varu: 160 °C (1,5 mmHg)
Storage Temperature: Refrigerator
MDL Number: MFCD00000414
CAS číslo: 23586-53-0
EINECS: 245-761-1
UN: 1707
ADR: 6.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR