Thallium(III) trifluoroacetate tech.
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Thallic trifluoroacetate
420760250
420761000
ACRO420760250EA
7030
CZK
ACRO420760250
ACRO420761000
Thallium(III) trifluoroacetate tech.
Thallium(III) trifluoroacetate
Vzorec:
C₆F₉O₆Tl Bod varu: 160 °C (1,5 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00000414 CAS číslo: 23586-53-0 EINECS: 245-761-1 UN: 1707 ADR: 6.1,II |
Výsledky specifikačního testu
Appearance | Light yellow to beige Powder |
Infrared spectrum | Conforms |
Melting point | 56 to 60 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...