Pentafluoropropionic acid, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Perfluoropropionic acid, Pentafluoropropanoic acid, Perfluoropropanoic acid, 2,2,3,3,3-Pentafluoropropionic acid
M-Clarity™ quality level = MQ100
245917-10G
245917-50G
SIAL245917-10GEA
1690
CZK
SIAL245917-10G
SIAL245917-50G
Pentafluoropropionic acid, Sigma-Aldrich®
Pentafluoropropionic acid
Vzorec:
C₃HF₅O₂ Bod varu: 96 °C (1013 hPa) Hustota: 1,561 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004170 CAS číslo: 422-64-0 EINECS: 207-021-6 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...