Kyselina heptafluorbutanová ≥99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Perfluorobutyric acid, HFBA, Heptafluorobutanoic acid
A14907.14
A14907.22
A14907.36
A14907.14EA
2080
CZK
A14907.14
A14907.22
A14907.36
Kyselina heptafluorbutanová ≥99%
Kyselina heptafluorbutanová
Purity: 99%
CAS No.: 375-22-4
Molecular Formula: C4HF7O2
Formula Weight: 214.04
Flash Point: None
Refractive Index: 1.300
Odour: Unpleasant
Melting Point: -18°
Boiling Point: 120-121°
Beilstein Reference: 1426882
UN No.: 3265
MDL No.: MFCD00004171
Vzorec:
C₄HF₇O₂ MW: 214,04 g/mol Bod varu: 120…121 °C Bod tání: –18 °C Hustota: 1,648 Storage Temperature: Ambient |
MDL Number:
MFCD00004171 CAS číslo: 375-22-4 EINECS: 206-786-3 UN: 3265 ADR: 8,II |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...