Ethyl trifluoroacetate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Trifluoroacetic ethyl ester
M-Clarity™ quality level = MQ200
E50000-100G
E50000-25G
SIALE50000-100GEA
3700
CZK
SIALE50000-100G
SIALE50000-25G
Ethyl trifluoroacetate, Sigma-Aldrich®
Ethyl trifluoroacetate
Vzorec:
C₄H₅F₃O₂ MW: 142,08 g/mol Bod varu: 61 °C (1013 hPa) Bod tání: –78 °C Hustota: 1,19 g/cm³ (20 °C) Bod vzplanutí: –17 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000419 CAS číslo: 383-63-1 EINECS: 206-851-6 UN: 2924 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...