Ethyl difluoroacetate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Difluoroacetic ethyl ester
M-Clarity™ quality level = MQ100
287628-25G
287628-5G
SIAL287628-25GEA
9000
CZK
SIAL287628-25G
SIAL287628-5G
Ethyl difluoroacetate, Sigma-Aldrich®
Ethyl difluoroacetate
Vzorec:
C₄H₆F₂O₂ MW: 124.09 Bod varu: 98 Hustota: 1.1893 g/cm³ (9.8 °C) |
MDL Number:
MFCD00013578 CAS číslo: 454-31-9 EINECS: 207-223-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...