Tisk…

Fluoroacetone, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

115460-1G 115460-5G
SIAL115460-1GEA 2740 CZK
SIAL115460-1G SIAL115460-5G
Fluoroacetone, Sigma-Aldrich®
Fluoroacetone
Vzorec: C₃H₅FO
Bod varu: 70…72 °C (1013 hPa)
Hustota: 1,054 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000451
CAS číslo: 430-51-3
EINECS: 207-064-0
UN: 2929
ADR: 6.1,I

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR