1,1,1-Trifluoroacetone, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
T62804-100G
T62804-25G
T62804-5G
SIALT62804-100GEA
10160
CZK
SIALT62804-100G
SIALT62804-25G
SIALT62804-5G
1,1,1-Trifluoroacetone, Sigma-Aldrich®
1,1,1-Trifluoroacetone
Vzorec:
C₃H₃F₃O Bod varu: 22 °C (1013 hPa) Bod tání: –78 °C Hustota: 1,18 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000423 CAS číslo: 421-50-1 EINECS: 207-005-9 UN: 1993 ADR: 3,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...