Tisk…

thioacetamid, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Ethanethioamide

M-Clarity™ quality level = MQ200

163678-100G 163678-25G 163678-500G
SIAL163678-100GEA 4460 CZK
SIAL163678-100G SIAL163678-25G SIAL163678-500G
thioacetamid, Sigma-Aldrich®
thioacetamid
Vzorec: CH₃CSNH₂
MW: 75,13 g/mol
Bod tání: 113…114 °C
Hustota: 1,336 g/cm³ (20 °C)
MDL Number: MFCD00008070
CAS číslo: 62-55-5
EINECS: 200-541-4
Merck Index: 13,09391
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe2000e62db991c85340c257ac6d64ca42c3551a&vid=18e76b2cdee9ee22a2618ec371858a498f218457&prid=web&clid=SAPDAM

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR