thioacetamid ≥99.0%, crystalline powder ACS
Dodavatel: MP Biomedicals
|
Nebezpečí
|
Synonyms:
Ethanethioamide
0215258780
0215258790
0215258725
ICNA0215258780EA
3800
CZK
ICNA0215258780
ICNA0215258790
ICNA0215258725
thioacetamid ≥99.0%, crystalline powder ACS
thioacetamid
Thioacetamide is a hepatotoxin and a carcinogen. Thioacetamide is also an additive in enantioselective reduction of β-keto esters with immobilized bakers yeast.
Thioacetamide is a source of hydrogen sulphide in qualitative analysis. It is a potent hepatotoxicant.
Vzorec:
CH₃CSNH₂ MW: 75,13 g/mol Bod tání: 111…114 °C Hustota: 1,336 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008070 CAS číslo: 62-55-5 EINECS: 200-541-4 Merck Index: 13,09391 |
Výsledky specifikačního testu
Assay | >99,0% |
Melting Point | 111 - 114°C |
Residue on Ignition | <0,05% |
Clarity of a 2% Solution | Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...