Tisk…

thioacetamid ≥99% ACS

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Ethanethioamide

30638.09 30638.18 30638.30
30638.09EA 1480 CZK
30638.09 30638.18 30638.30
thioacetamid ≥99% ACS
thioacetamid

Purity: 99% CAS No.: 62-55-5 Molecular Formula: CH3CSNH2 Formula Weight: 75.13 Physical Form: Crystalline Melting Point: 112-114° Beilstein Reference: 506006 Merck Reference: 14,9319 MDL No.: MFCD00008070

Vzorec: CH₃CSNH₂
MW: 75,13 g/mol
Bod tání: 112…114 °C
Hustota: 1,336 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008070
CAS číslo: 62-55-5
EINECS: 200-541-4
Merck Index: 14,9319
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe2000e62db991c85340c257ac6d64ca42c3551a&vid=18e76b2cdee9ee22a2618ec371858a498f218457&prid=web&clid=SAPDAM

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR