thioacetamid 99+% ACS
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Ethanethioamide
424530250
424531000
424535000
ACRO424530250EA
1800
CZK
ACRO424530250
ACRO424531000
ACRO424535000
thioacetamid 99+% ACS
thioacetamid
Vzorec:
CH₃CSNH₂ MW: 75,13 g/mol Bod tání: 112…114 °C Hustota: 1,37 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00008070 CAS číslo: 62-55-5 EINECS: 200-541-4 Merck Index: 13,09391 |
Výsledky specifikačního testu
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 111 to 114 °C |
Titration Argentometric | ≥99.0 % |
Residue after ignition | ≤0.05 % |
Clarity of solution | (2% soln.) Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...