Tisk…

Phenyl acetate, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: Acetic acid phenyl ester

M-Clarity™ quality level = MQ100

108723-100G 108723-5G
SIAL108723-100GEA 829 CZK
SIAL108723-100G SIAL108723-5G
Phenyl acetate, Sigma-Aldrich®
Phenyl acetate
Vzorec: C₈H₈O₂
MW: 136.15
Bod varu: 196
Hustota: 1.08 g/cm³ (20 °C)
Bod vzplanutí: 79
Storage Temperature: Ambient
MDL Number: MFCD00008699
CAS číslo: 122-79-2
EINECS: 204-575-0
Merck Index: 12,07421

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR