Tisk…

Hydrogencitran diamonný, Sigma-Aldrich®

Dodavatel: Merck
Varování


M-Clarity™ quality level = MQ200

09833-100G 09833-500G
SIAL09833-100GEA 3420 CZK
SIAL09833-100G SIAL09833-500G
Hydrogencitran diamonný, Sigma-Aldrich®
Hydrogencitran diamonný
Vzorec: HOC(CO₂H)(CH₂CO₂NH₄)₂
Hustota: 1,483 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013068
CAS číslo: 3012-65-5
EINECS: 221-146-3
Merck Index: 13,00513

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR