Trisodium citrate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Sodium Citrate, Sodium citrate tribasic, Citric acid trisodium salt, Sodium 2-hydroxypropane-1,2,3-tricarboxylate
C3674-100G
C3674-500G
C3674-1KG
SIALC3674-100GEA
1610
CZK
SIALC3674-100G
SIALC3674-500G
SIALC3674-1KG
Trisodium citrate, Sigma-Aldrich®
Trisodium citrate
Vzorec:
C₆H₅Na₃O₇ Bod tání: 150 °C Hustota: 1,815 g/cm³ (20 °C) Bod vzplanutí: 290 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012462 CAS číslo: 68-04-2 EINECS: 200-675-3 Merck Index: 12,08746 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...