Azelaic acid ≥96%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Azelaic acid,, 1,7-Heptanedicarboxylic acid, 1,9-Nonanedioic acid
42587.22
42587.36
42587.22EA
2290
CZK
42587.22
42587.36
Azelaic acid ≥96%
Azelaic acid
Purity: 96%
CAS No.: 123-99-9
Molecular Formula: HOOC(CH2)7COOH
Formula Weight: 188.22
Flash Point: 215°(419°F)
Physical Form: Crystalline
Density: 1.225
Melting Point: ca 102°
Boiling Point: 286°/100mm
Beilstein Reference: 1101094
Merck Reference: 14,905
MDL No.: MFCD00004432
Vzorec:
C₉H₁₆O₄ MW: 188,22 g/mol Bod varu: 286 °C (100 mmHg) Bod tání: ∼ 102 °C Hustota: 1,225 Bod vzplanutí: 215 °C (419 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004432 CAS číslo: 123-99-9 EINECS: 204-669-1 Merck Index: 14,905 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...