Tisk…

Suberic acid ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

A13963.22 A13963.0E A13963.36
A13963.22EA 1120 CZK
A13963.22 A13963.0E A13963.36
Suberic acid ≥99%
Suberic acid

Purity: 99% CAS No.: 505-48-6 Molecular Formula: HO2C(CH2)6CO2H Formula Weight: 174.20 Flash Point: 203°(397°F) Melting Point: 141-144° Boiling Point: 230°/15mm Beilstein Reference: 1210161 Merck Reference: 14,8862 MDL No.: MFCD00004428

Vzorec: C₈H₁₄O₄
Bod varu: 230 °C (15 mmHg)
Bod tání: 141…144 °C
Hustota: 1,354 g/cm³ (21 °C)
Bod vzplanutí: 203 °C (397 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004428
CAS číslo: 505-48-6
EINECS: 208-010-9
Merck Index: 14,8862

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR