Kyselina adipová ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Hexanedioc acid, 1,6-Hexanedioic acid
A13705.0B
A13705.0G
A13705.0I
A13705.0BEA
1170
CZK
A13705.0B
A13705.0G
A13705.0I
Kyselina adipová ≥99%
Kyselina adipová
Purity: 99%
CAS No.: 124-04-9
Molecular Formula: HOOC(CH2)4COOH
Formula Weight: 146.14
Flash Point: 196°(384°F)
Odour: Odourless
pH: 2.7
Melting Point: 151-155°
Boiling Point: ca 205°/3mm
Beilstein Reference: 1209788
Merck Reference: 14,162
MDL No.: MFCD00004420
Vzorec:
HOOC(CH₂)₄COOH Bod varu: ∼205 °C (3 mmHg) Bod tání: 151…155 °C Hustota: 1,360 Bod vzplanutí: 196 °C (384 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004420 CAS číslo: 124-04-9 EINECS: 204-673-3 Merck Index: 14,162 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...