1,16-Hexadecanedioic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
1,14-Tetradecanedicarboxylic acid, Thapsic acid, Hexadecanedioic acid
M-Clarity™ quality level = MQ200
177504-1G
177504-5G
SIAL177504-1GEA
2510
CZK
SIAL177504-1G
SIAL177504-5G
1,16-Hexadecanedioic acid, Sigma-Aldrich®
1,16-Hexadecanedioic acid
Vzorec:
C₁₆H₃₀O₄ Bod varu: 430 °C (1013 hPa) Bod tání: 122…123 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002746 CAS číslo: 505-54-4 EINECS: 208-013-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...