Kyselina sebaková ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Decanedioic acid, 1,8-Octanedicarboxylic acid, 1,10-Decanedioic acid
A14158.0B
A14158.0I
A14158.22
A14158.30
A14158.0BEA
4710
CZK
A14158.0B
A14158.0I
A14158.22
A14158.30
Kyselina sebaková ≥98%
Kyselina sebaková
Purity: 98+%
CAS No.: 111-20-6
Molecular Formula: C10H18O4
Formula Weight: 202.25
Flash Point: 220°(428°F)
Melting Point: 131-134°
Boiling Point: 295°/100mm
Beilstein Reference: 1210591
Merck Reference: 14,8415
MDL No.: MFCD00004440
Vzorec:
C₁₀H₁₈O₄ MW: 202,25 g/mol Bod varu: 295 °C (100 mmHg) Bod tání: 131…134 °C Hustota: 1,271 Bod vzplanutí: 220 °C (428 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004440 CAS číslo: 111-20-6 EINECS: 203-845-5 REACH: 01-2119519212-52 Merck Index: 14,8415 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...