Kyselina thioglykolová ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Mercaptoacetic acid
B20391.0E
B20391.22
B20391.36
B20391.C6
B20391.0EEA
8510
CZK
B20391.0E
B20391.22
B20391.36
B20391.C6
Kyselina thioglykolová ≥97%
Kyselina thioglykolová
Purity: 97+%
CAS No.: 68-11-1
Molecular Formula: C2H4O2S
Formula Weight: 92.11
Flash Point: 119°(246°F)
Physical Form: Clear liquid
Density: 1.295
Refractive Index: 1.5045
Sensitivity: Air Sensitive
Appearance: Colourless
Odour: Stench
Melting Point: -12°
Boiling Point: 220°
Beilstein Reference: 506166
Merck Reference: 14,9336
UN No.: 1940
MDL No.: MFCD00004876
Reagent that protects tryptophan in amino acid analysis
Vzorec:
HSCH₂COOH MW: 92,12 g/mol Bod varu: 220 °C Bod tání: –12 °C Hustota: 1,295 Bod vzplanutí: 119 °C (246 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004876 CAS číslo: 68-11-1 EINECS: 200-677-4 UN: 1940 ADR: 8,II Merck Index: 14,9336 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...