Tisk…

Kyselina glykolová ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 2-Hydroxy acid, 2-Hydroxyacetic acid

A12511.22 A12511.36
A12511.22EA 2030 CZK
A12511.22 A12511.36
Kyselina glykolová ≥98%
Kyselina glykolová

Purity: 98% CAS No.: 79-14-1 Molecular Formula: C2H4O3 Formula Weight: 76.05 Physical Form: Crystalline powder Density: 1.49 Appearance: White Odour: Unpleasant pH: 2 Melting Point: 74-80° Beilstein Reference: 1209322 Merck Reference: 14,4498 UN No.: 3261 MDL No.: MFCD00004312

Vzorec: C₂H₄O₃
Bod varu: 266 °C (1013 hPa)
Bod tání: 74…80 °C
Hustota: 1,49
Bod vzplanutí: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00004312
CAS číslo: 79-14-1
EINECS: 201-180-5
UN: 3261
ADR: 8,II
Merck Index: 14,4498

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR