Tisk…

12-Hydroxylauric acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 12-Hydroxydodecanoic acid, 12-Hydroxydodecylic acid

198781-1G 198781-25G 198781-5G
SIAL198781-1GEA 2160 CZK
SIAL198781-1G SIAL198781-25G SIAL198781-5G
12-Hydroxylauric acid, Sigma-Aldrich®
12-Hydroxylauric acid
Vzorec: C₁₂H₂₄O₃
MDL Number: MFCD00002739
CAS číslo: 505-95-3
EINECS: 208-025-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR