Tisk…
L-Methionin, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: (S)-(-)-2-Amino-4-(methylthio)butyric acid, (S)-(-)-Methionine, L(-)-Methionine, (S)-(-)-2-Amino-4-(methylthio)butanoic acid, H-Met-OH, Methionine, (S)-2-Amino-4-(methylthio)butanoic acid
M-Clarity™ quality level = MQ200
M5308-100G M5308-10MG M5308-1KG M5308-25G
SIALM5308-100GEA 9050 CZK
SIALM5308-100G SIALM5308-10MG SIALM5308-1KG SIALM5308-25G
L-Methionin, Sigma-Aldrich®
L-Methionin
Vzorec: C₅H₁₁NO₂S
MW: 149.21
Bod varu: 278
Bod tání: 136
Hustota: 1.373
Storage Temperature: Ambient
MDL Number: MFCD00063097
CAS číslo: 63-68-3
EINECS: 200-562-9
Merck Index: 13,06004