Tisk…

D-Methionine ≥99%

Dodavatel: Thermo Fisher Scientific

Synonyms: D(+)-Methionine, (R)-2-Amino-4-(methylthio)butanoic acid, (R)-(+)-2-Amino-4-(methylthio)butyric acid, (R)-(+)-2-Amino-4-(methylthio)butanoic acid, (R)-(+)-Methionine, H-D-Met-OH

B21213.06 B21213.14 B21213.22
B21213.06EA 958 CZK
B21213.06 B21213.14 B21213.22
D-Methionine ≥99%
D-Methionine

Purity: 99% CAS No.: 348-67-4 Molecular Formula: C5H11NO2S Formula Weight: 149.21 Melting Point: ca 280° dec. Beilstein Reference: 1722293 Merck Reference: 14,5975 Optical Rotation: -23° (c=2 in 2N HCl) MDL No.: MFCD00002622

Vzorec: C₅H₁₁NO₂S
Bod varu: 278 °C (1013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00002622
CAS číslo: 348-67-4
EINECS: 206-483-6
Merck Index: 14,5975

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR