Glycin, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Aminoacetic acid, Glycocoll, H-Gly-OH
M-Clarity™ quality level = MQ300
W328707-10KG
W328707-1KG
W328707-25KG
W328707-SAMPLE
SIALW328707-10KGEA
10600
CZK
SIALW328707-10KG
SIALW328707-1KG
SIALW328707-25KG
SIALW328707-SAMPLE
Glycin, Sigma-Aldrich®
Glycin
Vzorec:
NH₂CH₂COOH Bod varu: 181 °C (1013 hPa) Bod tání: 83 °C Hustota: 1,64 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008131 CAS číslo: 56-40-6 EINECS: 200-272-2 Merck Index: 13,04504 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...