Tisk…

Glycin, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

G8790-100G G8790-1KG
SIALG8790-100GEA 2740 CZK
SIALG8790-100G SIALG8790-1KG
Glycin, Sigma-Aldrich®
Glycin
Vzorec: NH₂CH₂COOH
Bod varu: 181 °C (1013 hPa)
Bod tání: 83 °C
Hustota: 1,64 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008131
CAS číslo: 56-40-6
EINECS: 200-272-2
Merck Index: 13,04504
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR