Tisk…

Manganese(II) acetate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Acetic acid manganese (II) salt, Manganese diacetate

M-Clarity™ quality level = MQ100

330825-100G 330825-25G 330825-5G
SIAL330825-100GEA 10370 CZK
SIAL330825-100G SIAL330825-25G SIAL330825-5G
Manganese(II) acetate, Sigma-Aldrich®
Manganese(II) acetate
Vzorec: (H₃CCOO)₂Mn
MW: 173.03
Storage Temperature: Ambient
MDL Number: MFCD00013039
CAS číslo: 638-38-0
EINECS: 211-334-3
Merck Index: 12,05763

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR