Tisk…

Stearan zinečnatý (12,5 - 14% ZnO)

Dodavatel: Thermo Scientific

Synonyms: Stearic acid zinc salt, Zinc(II) stearate

33238.A1 33238.A7 33238.30 33238.18
33238.A1EA 1860 CZK
33238.A1 33238.A7 33238.30 33238.18
Stearan zinečnatý (12,5 - 14% ZnO)
Stearan zinečnatý

CAS No.: 557-05-1 Molecular Formula: Zn[CH3(CH2)16CO2]2 Formula Weight: 632.48 Physical Form: Powder Appearance: White Melting Point: 130° MDL No.: MFCD00013031

Vzorec: C₃₆H₇₀O₄Zn
MW: 632,34 g/mol
Bod tání: 130 °C
Hustota: 1,095 g/cm³ (20 °C)
Bod vzplanutí: 276 °C
Storage Temperature: Ambient
MDL Number: MFCD00013031
CAS číslo: 557-05-1
EINECS: 209-151-9
Merck Index: 14,10158
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8ad7f3e101681798edbd54e57ad5d2ac82b2e5d&vid=435308cdcb35862fe6cc06384da2ef10d84c5700&prid=web&clid=SAPDAM

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR