Zinc acetate, anhydrous ≥99.98% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Varování
|
35792.09
35792.18
AB206958.0010
35792.09EA
2520
CZK
35792.09
35792.18
ABCRAB206958.0010
Zinc acetate, anhydrous ≥99.98% (metals basis)
Zinc acetate
Purity: 99.98%
CAS No.: 557-34-6
Molecular Formula: C4H6O4Zn
Formula Weight: 183.47
Physical Form: Granular
Density: 1.84
Appearance: White
Melting Point: 200° dec.
Merck Reference: 14,10128
UN No.: 3077
MDL No.: MFCD00012454
Vzorec:
C₄H₆O₄Zn Bod varu: 908 °C (1013 hPa) Hustota: 1,84 Storage Temperature: Ambient |
MDL Number:
MFCD00012454 CAS číslo: 557-34-6 EINECS: 209-170-2 UN: 3077 ADR: 9,III Merck Index: 14,10128 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...