Octan rtuťnatý ≥98.0% ACS
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Mercury diacetate, Mercuric acetate, Acetic acid mercury (II) salt
12273.18
12273.30
12273.18EA
1540
CZK
12273.18
12273.30
Octan rtuťnatý ≥98.0% ACS
Octan rtuťnatý
Purity: 98.0% min
CAS No.: 1600-27-7
Molecular Formula: C4H6HgO4
Formula Weight: 318.68
Physical Form: Crystalline
Density: 3.28
Appearance: White
Odour: Weak
Melting Point: 179-182°
Beilstein Reference: 3563831
Merck Reference: 14,5873
UN No.: 1629
MDL No.: MFCD00012165
Vzorec:
(H₃CCOO)₂Hg MW: 318.68 Bod tání: 179…182 °C Hustota: 3.27 Storage Temperature: Ambient |
MDL Number:
MFCD00012165 CAS číslo: 1600-27-7 EINECS: 216-491-1 UN: 1629 ADR: 6.1 II Merck Index: 14,5873 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...