Copper(II) acetate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Acetic acid copper (II) salt, Cupric acetate, Copper diacetate
M-Clarity™ quality level = MQ100
517453-25G
517453-5G
SIAL517453-25GEA
15220
CZK
SIAL517453-25G
SIAL517453-5G
Copper(II) acetate, Sigma-Aldrich®
Copper(II) acetate
Vzorec:
CuCH₃CO₂ MW: 181.64 Bod varu: 240 Bod tání: 115 Hustota: 1.9 Storage Temperature: Ambient |
MDL Number:
MFCD00008690 CAS číslo: 142-71-2 EINECS: 205-553-3 UN: 3077 ADR: 9 III Merck Index: 12,02690 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...