Copper(II) acetate, anhydrous
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Acetic acid copper (II) salt, Cupric acetate, Copper diacetate
19417.14
19417.22
19417.36
19417.14EA
3610
CZK
19417.14
19417.22
19417.36
Copper(II) acetate, anhydrous
Copper(II) acetate
CAS No.: 142-71-2
Molecular Formula: Cu(OOCCH3)2
Formula Weight: 181.63
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: Blue-green
Odour: Odourless
Merck Reference: 14,2624
UN No.: 3077
MDL No.: MFCD00008690
Vzorec:
CuCH₃CO₂ Bod varu: 240 °C (1013 hPa) Bod tání: 115 °C Hustota: 1,9 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008690 CAS číslo: 142-71-2 EINECS: 205-553-3 UN: 3077 ADR: 9,III Merck Index: 14,2624 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...