N-Lauroylsarcosine sodná sůl, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Lauroylsarcosine sodium, Sarkosyl, Sodium lauroyl sarcosine, Sodium N-lauroylsarcosinate, Sodium N-dodecanoyl-N-methylglycinate
61745-1KG
61745-250G
61745-50G
SIAL61745-1KGEA
8920
CZK
SIAL61745-1KG
SIAL61745-250G
SIAL61745-50G
N-Lauroylsarcosine sodná sůl, Sigma-Aldrich®
N-Lauroylsarcosine sodná sůl
Vzorec:
H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COONa MW: 293,38 g/mol Bod tání: 45…47 °C Bod vzplanutí: >150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042728 CAS číslo: 137-16-6 EINECS: 205-281-5 UN: 2811 ADR: 6.1,II Merck Index: 12,04379 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...