N-Lauroylsarcosine sodná sůl, Reagent Grade
Dodavatel: VWR Chemicals
|
Nebezpečí
|
Synonyms:
Lauroylsarcosine sodium, Sarkosyl, Sodium lauroyl sarcosine, Sodium N-lauroylsarcosinate, Sodium N-dodecanoyl-N-methylglycinate
0719-1KGEA
12890
CZK
0719-1KG
0719-500G
N-Lauroylsarcosine sodná sůl, Reagent Grade
N-Lauroylsarcosine sodná sůl
Anionic - negatively-charged (carboxylates, sulfates or sulfonates in alkali metal form). Utilised for applications ranging from solubilising membrane proteins in non-denaturing conditions to creating denaturing conditions during gel electrophoresis.
- Very fine white crystalline powder
Vzorec:
H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COONa MW: 293,38 g/mol Bod tání: 45…47 °C Bod vzplanutí: >150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042728 CAS číslo: 137-16-6 EINECS: 205-281-5 UN: 2811 ADR: 6.1,II Merck Index: 12,04379 |
Výsledky specifikačního testu
pH (10%, Water) @25 °C | 7.5 – 8.5 |
Purity | >95.0 % |
Sodium Laurate | ≤ 4.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...