Antimony triacetate ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Antimony acetate, Anitmony (III) acetate, Triacetoxystibine, Antimony(III) acetate, Antimony triacetate, Acetic acid antimony (III) salt
87657.22
87657.36
87657.09
87657.A3
87657.22EA
1610
CZK
87657.22
87657.36
87657.09
87657.A3
Antimony triacetate ≥97%
Antimony triacetate
Purity: 97%
CAS No.: 6923-52-0
Molecular Formula: Sb(OOCCH3)3
Formula Weight: 298.88
Physical Form: Powder
Sensitivity: Moisture Sensitive
Solubility: Decomposes in water
UN No.: 1549
MDL No.: MFCD00014974
Vzorec:
(H₃CCOO)₃Sb MW: 298.89 Storage Temperature: Ambient |
MDL Number:
MFCD00014974 CAS číslo: 6923-52-0 EINECS: 230-043-2 UN: 1549 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...